Learn More
Strem, An Ascensus Company CAS# 1643-19-2. 100g. Tetrabutylammonium bromide, 99%. MFCD00011633
SDP
Catalog No. 5090108602
Encompass
CAS# 1643-19-2. 100g. Tetrabutylammonium bromide, 99%. MFCD00011633. Molecular Weight: 322.37. Molecular Formula: (C4H9)4NBr. Color/form: white xtl. Strem# 02-5005. http://www.strem.com/catalog/v/02-5005/
Catalog No. 50-901-08602 Supplier Strem, An Ascensus Company Supplier No. 025005100G
CAS# 1643-19-2. 100g. Tetrabutylammonium bromide, 99%. MFCD00011633. Molecular Weight: 322.37. Molecular Formula: (C4H9)4NBr. Color/form: white xtl. Strem# 02-5005. http://www.strem.com/catalog/v/02-5005/