Learn More
Medchemexpress LLC Tetrabutylammonium bromide | 1643-19-2 | ≥98.0% | 1 KG
SDP
Catalog No. 5000410645
Encompass_Preferred
Tetrabutylammonium bromide is an organic ammonium compound frequently utilized in catalytic reactions and separation/purification processes. It exhibits a notable catalytic effect in certain organic synthesis reactions and functions as a surfactant, stabilizer, and antibacterial agent. Additionally, in laboratory research, this compound has been employed as an ion exchanger and solvent extractant.
  • Used in catalytic reactions.
  • Aids in separation and purification processes.
  • Provides significant catalytic effect in some organic synthesis reactions.
  • Can serve as a surfactant.
  • Functions as a stabilizer.
  • Acts as an antibacterial agent.
  • Utilized as an ion exchanger in laboratory studies.
  • Employed as a solvent extractant in laboratory studies.
  • Biochemical reagent for life science related research.
Catalog No. 50-004-10645 Supplier Medchemexpress LLC Supplier No. HYD01721KG
Tetrabutylammonium bromide is an organic ammonium compound frequently utilized in catalytic reactions and separation/purification processes. It exhibits a notable catalytic effect in certain organic synthesis reactions and functions as a surfactant, stabilizer, and antibacterial agent. Additionally, in laboratory research, this compound has been employed as an ion exchanger and solvent extractant.
  • Used in catalytic reactions.
  • Aids in separation and purification processes.
  • Provides significant catalytic effect in some organic synthesis reactions.
  • Can serve as a surfactant.
  • Functions as a stabilizer.
  • Acts as an antibacterial agent.
  • Utilized as an ion exchanger in laboratory studies.
  • Employed as a solvent extractant in laboratory studies.
  • Biochemical reagent for life science related research.