Learn More
Medchemexpress LLC Tetrabutylammonium bromide | 1643-19-2 | ≥98.0% | 5 KG
SDP
Catalog No. 5000410665
Encompass_Preferred
Tetrabutylammonium bromide is an organic ammonium compound often used in catalytic reactions and separation and purification processes. It exhibits significant catalytic effects in some organic synthesis reactions and can function as a surfactant, stabilizer, and antibacterial agent. In laboratory studies, it has also been used as an ion exchanger and solvent extractant. It serves as a biochemical reagent for life science-related research.
  • Can be used as a biological material
  • Can be used as an organic compound for life science related research
  • Used in catalytic reactions
  • Used in separation and purification processes
  • Functions as a surfactant
  • Functions as a stabilizer
  • Functions as an antibacterial agent
  • Can be used as an ion exchanger
  • Can be used as a solvent extractant
Catalog No. 50-004-10665 Supplier Medchemexpress LLC Supplier No. HYD01725KG
Tetrabutylammonium bromide is an organic ammonium compound often used in catalytic reactions and separation and purification processes. It exhibits significant catalytic effects in some organic synthesis reactions and can function as a surfactant, stabilizer, and antibacterial agent. In laboratory studies, it has also been used as an ion exchanger and solvent extractant. It serves as a biochemical reagent for life science-related research.
  • Can be used as a biological material
  • Can be used as an organic compound for life science related research
  • Used in catalytic reactions
  • Used in separation and purification processes
  • Functions as a surfactant
  • Functions as a stabilizer
  • Functions as an antibacterial agent
  • Can be used as an ion exchanger
  • Can be used as a solvent extractant