Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium bromide, For ion pair chromatography, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111013301 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3301 50 g
11-101-3300 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3301 Supplier MilliporeSigma Supelco Supplier No. 8685750GF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00011633

Specifications

Melting Point 102°C to 106°C; 102°C to 106°C
CAS 1643-19-2
Linear Formula (CH3 CH2 CH2 CH2)4 N(Br)
Quantity 50 g
Molecular Weight (g/mol) 322.37
Formula Weight 322.37
Percent Purity ≥99.0% (AT); ≥99.0%
Grade For ion pair chromatography
Physical Form Crystals
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.