Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium chloride, For ion pair chromatography, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111012468 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2468 50 g
11-101-2467 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2468 Supplier MilliporeSigma Supelco Supplier No. 8685250GF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetrabutylammonium chloride (TBACl) is a quaternary ammonium salt.g

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00011635

Specifications

CAS 1112-67-0
Flash Point 113°C - Closed Cup
Linear Formula [CH3(CH2)3]4 NCl
Quantity 50 g
Molecular Weight (g/mol) 277.92
Formula Weight 277.92
Percent Purity ≥99.0% (AT); ≥99.0%
Grade For ion pair chromatography
Physical Form Crystals
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.