Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Dibromoaurate, TCI America™
SDP

Catalog No. T12611G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T12611G Supplier TCI America Supplier No. T12611G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50481-01-1
Molecular Formula C16H36AuBr2N
Molecular Weight (g/mol) 599.246
MDL Number MFCD00059119
InChI Key JHKGBGGRJMVGSW-UHFFFAOYSA-L
PubChem CID 44630509
IUPAC Name dibromogold(1-);tetrabutylazanium
SMILES CCCC[N+](CCCC)(CCCC)CCCC.Br[Au-]Br

Specifications

Quantity 1 g
Formula Weight 599.25
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Dibromoaurate
EINECSNumber (2)-0186
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.