Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Dichloroaurate, TCI America™
SDP

Catalog No. T1273100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1273100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1273100MG Supplier TCI America Supplier No. T1273100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50480-99-4
Molecular Formula C16H36AuCl2N
Molecular Weight (g/mol) 510.338
MDL Number MFCD00059125
InChI Key XZONJCOZEGOEDY-UHFFFAOYSA-L
PubChem CID 11071265
IUPAC Name dichlorogold(1-);tetrabutylazanium
SMILES CCCC[N+](CCCC)(CCCC)CCCC.Cl[Au-]Cl

Specifications

Melting Point 83°C
Quantity 100 mg
Formula Weight 510.34
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Dichloroaurate
EINECSNumber (2)-0186
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.