Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Dichlorobromide 97.0+%, TCI America™
SDP

Catalog No. T118625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T118625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T118625G Supplier TCI America Supplier No. T118625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13053-75-3
Molecular Formula C16H36BrCl2N
Molecular Weight (g/mol) 393.28
MDL Number MFCD00060054
InChI Key XKVIJLXGNLMYCT-UHFFFAOYSA-N
PubChem CID 6399486
IUPAC Name dichlorobromanuide; tetrabutylazanium
SMILES Cl[Br-]Cl.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 65°C
Quantity 25 g
Formula Weight 393.28
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Dichlorobromide
EINECSNumber (2)-0186
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.