Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium difluorotriphenyl stannate, 95%, Thermo Scientific™

Catalog No. AC302590010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC302590010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC302590010 Supplier Thermo Scientific Chemicals Supplier No. 302590010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

 

C-X bond formation reagent, Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139353-88-1
Molecular Formula C34H51F2NSn
Molecular Weight (g/mol) 630.48
MDL Number MFCD00192465
InChI Key ODMXVCNGZGLSRS-UHFFFAOYSA-L
Synonym tetrabutylammonium difluorotriphenylstannate, tetrabutylammonium difluorotriphenylstannate iv, tetrabutylammonium difluorotriphenyltin, tetrabutylammonium triphenyldifluorotin, tetrabutylammoniumdifluorotriphenylstannate, tetrabutylam-monium triphenyldifluorostannate, tetrabutylammonium difluorotriphenyl stannate, tetra-n-butylammonium difluorotriphenylstannate, difluorotriphenylstannanuide; tetrabutylammonium, difluoro triphenyl stannanuide; tetrabutylammonium
PubChem CID 2733194
IUPAC Name difluoro(triphenyl)stannanuide;tetrabutylazanium
SMILES CCCC[N+](CCCC)(CCCC)CCCC.C1=CC=C(C=C1)[Sn-](C2=CC=CC=C2)(C3=CC=CC=C3)(F)F

Specifications

Melting Point 187°C to 193°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 1 g
Formula Weight 630.48
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium difluorotriphenyl stannate, 95%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.