Learn More
Medchemexpress LLC Tetrabutylammonium difluorotriphenylsilicate(IV) (TBAT) | 163931-61-1 | 539.86 | 100 G
SDP
Catalog No. 5000449084
Encompass_Preferred
Tetrabutylammonium difluorotriphenylsilicate(IV) (TBAT) is an organic compound that serves as a catalyst and surfactant. It is utilized in various organic synthesis reactions as a catalytic agent and also functions as a cleaning agent, lubricant, and stabilizer. The compound finds extensive application in industrial sectors, including plastic, rubber, and textile manufacturing processes.
  • Functions as a catalyst and surfactant.
  • Can be used as a cleaning agent, lubricant, and stabilizer.
  • Widely used in industrial fields such as plastic, rubber, and textile manufacturing processes.
  • Biochemical reagent for life science-related research.
Catalog No. 50-004-49084 Supplier Medchemexpress LLC Supplier No. HYW004963100G
Tetrabutylammonium difluorotriphenylsilicate(IV) (TBAT) is an organic compound that serves as a catalyst and surfactant. It is utilized in various organic synthesis reactions as a catalytic agent and also functions as a cleaning agent, lubricant, and stabilizer. The compound finds extensive application in industrial sectors, including plastic, rubber, and textile manufacturing processes.
  • Functions as a catalyst and surfactant.
  • Can be used as a cleaning agent, lubricant, and stabilizer.
  • Widely used in industrial fields such as plastic, rubber, and textile manufacturing processes.
  • Biochemical reagent for life science-related research.