Learn More
Medchemexpress LLC Tetrabutylammonium difluorotriphenylsilicate(IV) (TBAT) | 163931-61-1 | 539.86 | 50 G
SDP
Catalog No. 5000446059
Encompass_Preferred
Tetrabutylammonium difluorotriphenylsilicate(IV) (TBAT) is an organic compound commonly used as a catalyst and surfactant. It can play a catalytic role in some organic synthesis reactions, and can be used as a cleaning agent, lubricant, and stabilizer. The compound is also widely used in certain industrial fields, such as in plastic, rubber, and textile manufacturing processes. It is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Acts as a catalyst in organic synthesis reactions.
  • Functions as a cleaning agent, lubricant, and stabilizer.
  • Applied in industrial fields, including plastic, rubber, and textile manufacturing processes.
  • Suitable as a biochemical reagent for life science research.
Catalog No. 50-004-46059 Supplier Medchemexpress LLC Supplier No. HYW00496350G
Tetrabutylammonium difluorotriphenylsilicate(IV) (TBAT) is an organic compound commonly used as a catalyst and surfactant. It can play a catalytic role in some organic synthesis reactions, and can be used as a cleaning agent, lubricant, and stabilizer. The compound is also widely used in certain industrial fields, such as in plastic, rubber, and textile manufacturing processes. It is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Acts as a catalyst in organic synthesis reactions.
  • Functions as a cleaning agent, lubricant, and stabilizer.
  • Applied in industrial fields, including plastic, rubber, and textile manufacturing processes.
  • Suitable as a biochemical reagent for life science research.