Learn More
Medchemexpress LLC Tetrabutylammonium (dihydrogen trifluoride) | 99337-56-1 | MFCD00145365 | 98.0% | 301.47 g/mol | C16H38F3N | 1 G
SDP
Catalog No. 5000283094
Encompass_Preferred
Tetrabutylammonium (dihydrogen trifluoride) is a quaternary ammonium salt used as a soluble fluoride source and electrolyte for research applications. It is provided as a high-purity reagent intended for synthetic chemistry, electrochemical studies, and materials research; typical properties include molecular formula C16H38F3N and molecular weight 301.47 g/mol.
  • Provides a soluble source of fluoride for organic synthesis.
  • Acts as an electrolyte in electrochemical and battery research.
  • High purity supports reproducible experimental results.
  • Available in small lab-scale pack sizes for benchtop use.
  • Soluble in common organic solvents for straightforward handling.
Catalog No. 50-002-83094 Supplier Medchemexpress LLC Supplier No. HYW1276871G
Tetrabutylammonium (dihydrogen trifluoride) is a quaternary ammonium salt used as a soluble fluoride source and electrolyte for research applications. It is provided as a high-purity reagent intended for synthetic chemistry, electrochemical studies, and materials research; typical properties include molecular formula C16H38F3N and molecular weight 301.47 g/mol.
  • Provides a soluble source of fluoride for organic synthesis.
  • Acts as an electrolyte in electrochemical and battery research.
  • High purity supports reproducible experimental results.
  • Available in small lab-scale pack sizes for benchtop use.
  • Soluble in common organic solvents for straightforward handling.