Learn More
Medchemexpress LLC Tetrabutylammonium (dihydrogen trifluoride) | 99337-56-1 | MFCD00145365 | 98.0% | 301.47 g·mol⁻¹ | C16H38F3N | 5 G
SDP
Catalog No. 5000283635
Encompass_Preferred
Tetrabutylammonium (dihydrogen trifluoride) is a quaternary ammonium salt provided as a high-purity solid reagent for research applications. It is used as a convenient source of dihydrogen trifluoride/fluoride in synthetic chemistry and as an ionic component in specialized electrolyte systems; store sealed and protected from moisture.
Catalog No. 50-002-83635 Supplier Medchemexpress LLC Supplier No. HYW1276875G
Tetrabutylammonium (dihydrogen trifluoride) is a quaternary ammonium salt provided as a high-purity solid reagent for research applications. It is used as a convenient source of dihydrogen trifluoride/fluoride in synthetic chemistry and as an ionic component in specialized electrolyte systems; store sealed and protected from moisture.