Learn More
Medchemexpress LLC Tetrabutylammonium fluoride | 429-41-4 | MFCD00011747 | 261.46 | C16H36FN | 500mL
SDP
Catalog No. 5000267825
Encompass_Preferred
Tetrabutylammonium fluoride is an organic compound containing both ammonium and fluorine functional groups It is commonly used as a reagent in various chemical synthesis applications especially as a source of fluoride ions for nucleophilic reactions Tetrabutylammonium fluoride has several properties that make it suitable for these applications including its high solubility in polar solvents and its ability to selectively activate certain chemical bonds In addition it can be used as a catalyst for organic reactions and as an electrolyte for batteries
Catalog No. 50-002-67825 Supplier Medchemexpress LLC Supplier No. HYY0971500ML
Tetrabutylammonium fluoride is an organic compound containing both ammonium and fluorine functional groups It is commonly used as a reagent in various chemical synthesis applications especially as a source of fluoride ions for nucleophilic reactions Tetrabutylammonium fluoride has several properties that make it suitable for these applications including its high solubility in polar solvents and its ability to selectively activate certain chemical bonds In addition it can be used as a catalyst for organic reactions and as an electrolyte for batteries