Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrabutylammonium Fluoride Solution

Catalog No. 361399500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-139-9500G 500 g
36-139-9100G 100 g
36-139-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 36-139-9500G Supplier Sigma Aldrich Supplier No. 361399500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for preparation of:Triple monoamine reuptake inhibitors as a new generation of antidepressantsAlcohols via hydrolysis of alkyl silyl ethers neutral pH in mixed organic-aqueous buffered solutionsOligoribonucleotides with phosphonate-modified linkagesAryl alkyl alcohols via Nozaki-Hiyama allylation catalyzed by chiral bipyridyldiol ligands and chromium trichlorideConjugated dienoic acid esters using Suzuki coupling reactionsMacrocyclic o-aminobenzamide Hsp90 inhibitorwith antitumor activityPhospshoinositide 3-kinase (PI3K)/mammalian target of rapamycin (mTOR) dual inhibitorsAnti-diabetic polyacetylenic glucosides

Specifications

CAS 429-41-4
Density 0.953 g/mL at 25°C
Molecular Formula C16H36FN
Linear Formula [CH3(CH2)3]4NF
MDL Number MFCD00011747
Quantity 500 g
Synonym TBAF solution
Molecular Weight (g/mol) 261.46 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.