Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrabutylammonium Fluoride Solution

Catalog No. 21614325L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
50 mL
100 mL
500 mL
1 L
2.5 L
4 x 100 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-614-325L 2.5 L
21-614-3100ML 4 x 100 mL
50-308-1469 100 mL
21-614-35ML 5 mL
21-614-31L 1 L
21-614-350ML 50 mL
21-614-3500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Danger
Catalog No. 21-614-325L Supplier Sigma Aldrich Supplier No. 2161432.5L
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetrabutylammonium fluoride (TBAF) can be used as a reagent: For the deprotection of silyl and N-sulfonyl groups. In the fluorination reactions.To synthesize 2-substituted indoles by cyclization reaction of various 2-ethynylanilines with terminal alkynes using Pd catalyst. It can also be used as an activator in the synthesis of arylated or alkenylated alkynes by the coupling reaction of aryl and alkenyl halides with terminal alkynes in the presence of Pd catalyst.

Specifications

CAS 429-41-4
Density 0.903 g/mL at 25°C
Molecular Formula C16H36FN
Linear Formula [CH3(CH2)3]4NF
Quantity 2.5 L
MDL Number MFCD00011747
Synonym TBAF solution
Molecular Weight (g/mol) 261.46 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.