Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium fluoride trihydrate, 99%

Catalog No. AC221080500
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221080500 50 g Glass bottle
AC221080100 10 g Glass bottle
AC221082500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC221080500 Supplier Thermo Scientific Chemicals Supplier No. 221080500
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 87749-50-6 | C16H36FN·3H2O | 315.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87749-50-6
Molecular Formula C16H36FN·3H2O
Molecular Weight (g/mol) 315.51
MDL Number MFCD00149981
InChI Key VEPTXBCIDSFGBF-UHFFFAOYSA-M
Synonym tetrabutylammonium fluoride trihydrate, tetra-n-butylammonium fluoride trihydrate, tetrabutylazanium fluoride trihydrate, tetrabutylammonium trihydrate fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, trihydrate, tetrabutylammonium ion trihydrate fluoride, tetra-n-butylammonium fluoride hydrate, bu4nf trihydrate, pubchem2275, pubchem4057
PubChem CID 11726816
IUPAC Name tetrabutylazanium;fluoride;trihydrate
SMILES CCCC[N+](CCCC)(CCCC)CCCC.O.O.O.[F-]

Specifications

Melting Point 58°C to 60°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula [CH3(CH2)3]4NF·3H2O
Quantity 50 g
Fieser 04,477; 05,645; 07,353; 08,467; 09,444; 10,378; 11,499; 12,458; 14,293; 15,168; 17,321
Solubility Information Solubility in water: soluble
Formula Weight 315.51
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Tetrabutylammonium fluoride trihydrate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.