Learn More
Strem, An Ascensus Company CAS# 3109-63-5. 25g. Tetrabutylammonium hexafluorophosphate, 98%. MFCD00011748
SDP
Catalog No. 5090109411
Encompass
CAS# 3109-63-5. 25g. Tetrabutylammonium hexafluorophosphate, 98%. MFCD00011748. Molecular Weight: 387.44. Molecular Formula: (C4H9)4NPF6. Color/form: white xtl. Strem# 09-6634. http://www.strem.com/catalog/v/09-6634/
Catalog No. 50-901-09411 Supplier Strem, An Ascensus Company Supplier No. 09663425G
CAS# 3109-63-5. 25g. Tetrabutylammonium hexafluorophosphate, 98%. MFCD00011748. Molecular Weight: 387.44. Molecular Formula: (C4H9)4NPF6. Color/form: white xtl. Strem# 09-6634. http://www.strem.com/catalog/v/09-6634/