Learn More
Medchemexpress LLC Tetrabutylammonium hexafluorophosphate | 3109-63-5 | MFCD00011748 | 250g
SDP
Catalog No. 5000266137
Encompass_Preferred
Tetrabutylammonium hexafluorophosphate is an organic-inorganic compound commonly used as an ionic solvent and catalyst It can be used to catalyze or promote reactions in certain chemical reactions and is widely used in batteries solar cells and pigments In addition this compound is also widely used in organic synthesis and chemical analysis
Catalog No. 50-002-66137 Supplier Medchemexpress LLC Supplier No. HYY1699250G
Tetrabutylammonium hexafluorophosphate is an organic-inorganic compound commonly used as an ionic solvent and catalyst It can be used to catalyze or promote reactions in certain chemical reactions and is widely used in batteries solar cells and pigments In addition this compound is also widely used in organic synthesis and chemical analysis