Learn More
Medchemexpress LLC Tetrabutylammonium hydrogen sulfate | 32503-27-8 | MFCD00011637 | 99.7% | 339.53 g/mol | C16H37NO4S | 500g
SDP
Supplier:  Medchemexpress LLC HYY1045500G
Encompass_Preferred
Tetrabutylammonium hydrosulfate is a quaternary ammonium salt that is commonly used as a phase transfer catalyst acid catalyst and reagent in various chemical reactions especially in organic synthesis Tetrabutylammonium hydrosulfate has unique chemical properties that allow it to facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes
Catalog No. 50-002-69653