Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium hydrogen sulfate, 99.15%, MP Biomedicals™

Catalog No. ICN15210325 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15210325 25 g
ICN15210380 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15210325 Supplier MP Biomedicals, Inc Supplier No. 0215210325

Chemical Identifiers

CAS 136-47-0
Molecular Formula C15H25ClN2O2
Molecular Weight (g/mol) 300.827
InChI Key PPWHTZKZQNXVAE-UHFFFAOYSA-N
Synonym tetracaine hydrochloride, tetracaine hcl, butethanol, butylocaine, anethain, anethaine, curtacain, decicain, decicaine, dessicain
PubChem CID 8695
IUPAC Name 2-(dimethylamino)ethyl 4-(butylamino)benzoate;hydrochloride
SMILES CCCCNC1=CC=C(C=C1)C(=O)OCCN(C)C.Cl

Specifications

Assay Percent Range >97%
Quantity 25 g
Formula Weight 339.5
Percent Purity 0.9915
Physical Form Powder
Chemical Name or Material Tetrabutylammonium Hydrogen Sulfate
EINECSNumber 251-068-5
Recommended Storage Store at 2° to 8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.