Learn More
Medchemexpress LLC Tetrabutylammonium hydrogensulfate | 32503-27-8 | MFCD00011637 | 339.53 | C16H37NO4S | 1kg
SDP
Catalog No. 5000266733
Encompass_Preferred
Tetrabutylammonium hydrosulfate is a quaternary ammonium salt that is commonly used as a phase transfer catalyst acid catalyst and reagent in various chemical reactions especially in organic synthesis Tetrabutylammonium hydrosulfate has unique chemical properties that allow it to facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes
Catalog No. 50-002-66733 Supplier Medchemexpress LLC Supplier No. HYY10451KG
Tetrabutylammonium hydrosulfate is a quaternary ammonium salt that is commonly used as a phase transfer catalyst acid catalyst and reagent in various chemical reactions especially in organic synthesis Tetrabutylammonium hydrosulfate has unique chemical properties that allow it to facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes