Learn More
Strem, An Ascensus Company CAS# 2052-49-5. 25g. Tetrabutylammonium hydroxide, 40% in water. MFCD00009425
SDP
Catalog No. 5090109116
Encompass
CAS# 2052-49-5. 25g. Tetrabutylammonium hydroxide, 40% in water. MFCD00009425. Molecular Weight: 259.47. Molecular Formula: (C4H9)4NOH. Color/form: colorless liq. Strem# 07-1967. http://www.strem.com/catalog/v/07-1967/
Catalog No. 50-901-09116 Supplier Strem, An Ascensus Company Supplier No. 07196725G
CAS# 2052-49-5. 25g. Tetrabutylammonium hydroxide, 40% in water. MFCD00009425. Molecular Weight: 259.47. Molecular Formula: (C4H9)4NOH. Color/form: colorless liq. Strem# 07-1967. http://www.strem.com/catalog/v/07-1967/