Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrabutylammonium Hydroxide Solution

Catalog No. 86863100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-863-100ML 25 g
86-863-500ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 86-863-100ML Supplier Sigma Aldrich Supplier No. 86863100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The product is 54.0-56.0% solution of tetrabutylammonium hydroxide in H2O. Tetrabutylammonium hydroxide has been synthesized from tetrabutylammonium iodide. TBAOH dissolved in anhydrous isopropyl alcohol is widely employed as titrant for weak acids.Tetrabutylammonium hydroxide solution (TBAOH) may be used in the following studies:For the dissolution of 10 wt% cellulose under room temperature conditions.As a catalyst during the synthesis of 1,2,4-oxadiazoles.Preparation of TBAOH salts of fatty acids.

Specifications

CAS 39139-87-2
Molecular Formula C17H36N2O
Linear Formula (CH3CH2CH2CH2)4N(OCN)
MDL Number MFCD00043222
Quantity 25 g
Molecular Weight (g/mol) 284.48 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.