Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrabutylammonium Hydroxide Solution

Catalog No. 230189100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
250 mL
800 mL
1 L
4 x 100 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-018-9100ML 100 mL
23-018-9800ML 800 mL
23-018-91L 1 L
50-308-1480 4 x 100 mL
23-018-9250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Danger
Catalog No. 23-018-9100ML Supplier Sigma Aldrich Supplier No. 230189100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetrabutylammonium hydroxide is a quaternary ammonium salt mainly used as a strong base and phase-transfer catalyst.Tetrabutylammonium hydroxide solution (1M in methanol) may be used as a base to synthesize ring methyl and methylene deuteriated porphyrins and metalloporphyrins. It may also be used as an initiator of graft polymerization between pivalolactone and poly(ethylene-co-vinyl acetate-co-methacrylic acid).

Specifications

CAS 2052-49-5
Density 0.83 g/mL at 25°C
Molecular Formula C16H37NO
Refractive Index n20/D 1.3775
Linear Formula (CH3CH2CH2CH2)4N(OH)
Quantity 100 mL
MDL Number MFCD00009425
Molecular Weight (g/mol) 259.47 g/mol
Biological Activity Eyes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.