Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Hydroxide Solution, ∼40 in Water, For ion chromatography, MilliporeSigma™ Supelco™

Catalog No. 111018079 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8079 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8079 Supplier MilliporeSigma Supelco Supplier No. 86854500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetrabutylammonium hydroxide (TBAOH) is a quaternary ammonium salt,mainly used as a strong base and phase-transfer catalyst.

Reagent suitable for use in ion chromatography
TRUSTED_SUSTAINABILITY

Specifications

CAS 2052-49-5
Density 0.995 g/cm3
Linear Formula (CH3CH2CH2CH2)4 N(OH)
MDL Number MFCD00009425
Quantity 500 mL
Molecular Weight (g/mol) 259.47
Formula Weight 259.47
Grade For ion chromatography
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.