Learn More
Tetrabutylammonium hydroxide Solution, ∼40 in Water, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 2052-49-5 |
| Quantity | 2.5 L |
| Linear Formula | (CH3 CH2 CH2 CH2)4 N(OH) |
| Formula Weight | 259.47 |
| Grade | Ion Chromatography |
| Concentration or Composition (by Analyte or Components) | 40% aq. soln. |
| Density | 0.995 g/cm3 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.