Learn More
Thermo Scientific TETRABUTYLAMMONIUM HYDROXIDE,
Catalog No. NC9390323 Shop All Thermo Scientific Products
Encompass
Cation Regenerant Solution (2.06 M TBAOH), 100 mL
Catalog No. NC9390323 Supplier Thermo Scientific Supplier No. 057561
Cation Regenerant Solution (2.06 M TBAOH), 100 mL