Learn More
Medchemexpress LLC Tetrabutylammonium iodide | 311-28-4 | MFCD00011636 | ≥99.9% | 369.37 g/mol | C16H36IN | 100g
SDP
Catalog No. 5000269654
Encompass_Preferred
Tetrabutylammonium iodide is a quaternary ammonium salt commonly used as a phase transfer catalyst surfactant and stabilizer in various chemical reactions especially in organic synthesis Tetrabutylammonium iodide has unique chemical properties that facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes
Catalog No. 50-002-69654 Supplier Medchemexpress LLC Supplier No. HYY0791100G
Tetrabutylammonium iodide is a quaternary ammonium salt commonly used as a phase transfer catalyst surfactant and stabilizer in various chemical reactions especially in organic synthesis Tetrabutylammonium iodide has unique chemical properties that facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes