Learn More
Medchemexpress LLC Tetrabutylammonium iodide | 311-28-4 | MFCD00011636 | ≥99.9% | 369.37 g/mol | C16H36IN | 500g
SDP
Catalog No. 5000268673
Encompass_Preferred
Tetrabutylammonium iodide is a quaternary ammonium salt commonly used as a phase transfer catalyst surfactant and stabilizer in various chemical reactions especially in organic synthesis Tetrabutylammonium iodide has unique chemical properties that facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes
Catalog No. 50-002-68673 Supplier Medchemexpress LLC Supplier No. HYY0791500G
Tetrabutylammonium iodide is a quaternary ammonium salt commonly used as a phase transfer catalyst surfactant and stabilizer in various chemical reactions especially in organic synthesis Tetrabutylammonium iodide has unique chemical properties that facilitate the transfer of ions or molecules from one phase to another making it an important ingredient in a variety of industrial processes