Learn More
Medchemexpress LLC Tetrabutylammonium p 100mg
SDP
Catalog No. 5000205541
Encompass_Preferred
Tetrabutylammonium perrhenate is a quaternary ammonium salt belonging to the class of alkylammonium salts This compound is used as a catalyst in organic synthesis especially in the production of alkenes and alcohols Furthermore due to its ability to bind radioisotopes it is useful as a radiopharmaceutical in nuclear medicine Its unique chemical properties make it an essential ingredient in a variety of industrial processes including petrochemical refining and pharmaceuticals
Catalog No. 50-002-05541 Supplier Medchemexpress LLC Supplier No. HYW109978100MG
Tetrabutylammonium perrhenate is a quaternary ammonium salt belonging to the class of alkylammonium salts This compound is used as a catalyst in organic synthesis especially in the production of alkenes and alcohols Furthermore due to its ability to bind radioisotopes it is useful as a radiopharmaceutical in nuclear medicine Its unique chemical properties make it an essential ingredient in a variety of industrial processes including petrochemical refining and pharmaceuticals