Learn More
Medchemexpress LLC Tetrabutylammonium perchlorate | 1923-70-2 | MFCD00038722 | 98.0% | 341.91 g/mol | C16H36ClNO4 | 100 G
SDP
Catalog No. 5000283628
Encompass_Preferred
Tetrabutylammonium perchlorate is a quaternary ammonium salt supplied as a solid reagent (CAS 1923-70-2). It is commonly used as an electrolyte in electrochemistry, as an ion-pairing reagent in analytical chemistry, and as a reagent in organic synthesis and materials research.
  • High purity (98.0%).
  • Solid quaternary ammonium salt with good thermal stability.
  • Soluble in common organic solvents, suitable for electrochemical electrolytes.
  • Useful as a source of perchlorate anion for analytical methods.
  • Suitable for organic synthesis, ion-pairing, and materials applications.
Catalog No. 50-002-83628 Supplier Medchemexpress LLC Supplier No. HYW127680100G
Tetrabutylammonium perchlorate is a quaternary ammonium salt supplied as a solid reagent (CAS 1923-70-2). It is commonly used as an electrolyte in electrochemistry, as an ion-pairing reagent in analytical chemistry, and as a reagent in organic synthesis and materials research.
  • High purity (98.0%).
  • Solid quaternary ammonium salt with good thermal stability.
  • Soluble in common organic solvents, suitable for electrochemical electrolytes.
  • Useful as a source of perchlorate anion for analytical methods.
  • Suitable for organic synthesis, ion-pairing, and materials applications.