Learn More
Medchemexpress LLC Tetrabutylammonium perchlorate | 1923-70-2 | MFCD00038722 | 98.0% | 341.91 g/mol | C16H36ClNO4 | 25 G
SDP
Catalog No. 5000282667
Encompass_Preferred
Tetrabutylammonium perchlorate is an ionic organic salt commonly used as an electrolyte and as a perchlorate source in analytical and electrochemical research. It is supplied as a research-grade reagent with high purity for use in electrochemistry, chromatographic methods, and analytical method development.
  • High purity (98.0%).
  • Useful as an electrolyte for electrochemical applications.
  • Suitable as a reference standard in ion chromatography and analytical methods.
  • Stable when stored sealed and kept away from moisture.
  • Supplied in laboratory package sizes for convenient handling.
Catalog No. 50-002-82667 Supplier Medchemexpress LLC Supplier No. HYW12768025G
Tetrabutylammonium perchlorate is an ionic organic salt commonly used as an electrolyte and as a perchlorate source in analytical and electrochemical research. It is supplied as a research-grade reagent with high purity for use in electrochemistry, chromatographic methods, and analytical method development.
  • High purity (98.0%).
  • Useful as an electrolyte for electrochemical applications.
  • Suitable as a reference standard in ion chromatography and analytical methods.
  • Stable when stored sealed and kept away from moisture.
  • Supplied in laboratory package sizes for convenient handling.