Learn More
Tetrabutylammonium perchlorate, For electrochemical analysis, ≥99.0%, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 1923-70-2 |
| Melting Point | 211°C to 215°C |
| Linear Formula | (CH3 CH2 CH2 CH2)4 N(ClO4) |
| Quantity | 10 g |
| Solubility Information | Acetonitrile: 0.1 g/mL, clear, colorless |
| Molecular Weight (g/mol) | 341.91 |
| Formula Weight | 341.91 |
| Percent Purity | ≥99.0% (T); ≥99.0% |
| Grade | For electrochemical analysis |
| Physical Form | Powder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.