Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Perchlorate 98.0+%, TCI America™
SDP

Catalog No. T0836100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0836100G 100 g
T083625G 25 g
T0836500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0836100G Supplier TCI America Supplier No. T0836100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1923-70-2
Molecular Formula C16H36ClNO4
Molecular Weight (g/mol) 341.917
MDL Number MFCD00038722
InChI Key KBLZDCFTQSIIOH-UHFFFAOYSA-M
Synonym tetrabutylammonium perchlorate, 1-butanaminium, n,n,n-tributyl-, perchlorate, tetrabutyl ammonium perchlorate, tetra-n-butylammonium perchlorate, tetrabutylammonium perchlorat, 1-butanaminium, n,n,n-tributyl-, perchlorate 1:1, acmc-1bpvk, n,n,n-tributylbutan-1-aminium perchlorate, tetrabutylammoniumperchlorate, tetrabutylazanium perchlorate
PubChem CID 74723
IUPAC Name tetrabutylazanium;perchlorate
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[O-]Cl(=O)(=O)=O

Specifications

Color White
Quantity 100 g
UN Number 1479
Formula Weight 341.92
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Perchlorate
EINECSNumber (2)-0186
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.