Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium Phosphate, ∽97%, MP Biomedicals™

Catalog No. ICN19551301 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19551301 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19551301 Supplier MP Biomedicals, Inc Supplier No. 0219551301
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ion-associating reagent for HPLC

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5574-97-0
Molecular Formula C16H38NO4P
Molecular Weight (g/mol) 339.46
MDL Number MFCD00064526
InChI Key ARRNBPCNZJXHRJ-UHFFFAOYSA-M
Synonym tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion
PubChem CID 2735142
IUPAC Name tetrabutylazanium dihydrogen phosphate
SMILES OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Color White
Assay Percent Range Approx. 97%
Quantity 1 g
Formula Weight 339.46
Physical Form Powder
Chemical Name or Material Tetrabutylammonium phosphate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.