Learn More
Medchemexpress LLC Tetrabutylammonium t 1g
SDP
Catalog No. 5000206350
Encompass_Preferred
Tetrabutylammonium tetraphenylborate is a quaternary ammonium salt belonging to the class of alkylammonium salts This compound is commonly used as an ion-pairing reagent in chromatography and analytical chemistry facilitating the separation and identification of various organic compounds Its unique chemical properties make it an important ingredient in a variety of scientific applications including pharmaceuticals agrochemicals and materials science products
Catalog No. 50-002-06350 Supplier Medchemexpress LLC Supplier No. HYW1114361G
Tetrabutylammonium tetraphenylborate is a quaternary ammonium salt belonging to the class of alkylammonium salts This compound is commonly used as an ion-pairing reagent in chromatography and analytical chemistry facilitating the separation and identification of various organic compounds Its unique chemical properties make it an important ingredient in a variety of scientific applications including pharmaceuticals agrochemicals and materials science products