Learn More
Medchemexpress LLC Tetrabutylammonium t 5g
SDP
Catalog No. 5000206351
Encompass_Preferred
Tetrabutylammonium thiocyanateIt is a quaternary ammonium salt and belongs to the category of alkyl ammonium salts This compound is widely used as a reagent in organic chemistry especially for the synthesis of various organic compounds including pharmaceuticals and agrochemicals also Tetrabutylammonium thiocyanateIt can also be used as an electrolyte in electrochemical devices and as a phase transfer catalyst in organic synthesis Its unique chemical properties make it an important ingredient in a variety of industrial processes including petrochemical refining and materials science
Catalog No. 50-002-06351 Supplier Medchemexpress LLC Supplier No. HYW1114615G
Tetrabutylammonium thiocyanateIt is a quaternary ammonium salt and belongs to the category of alkyl ammonium salts This compound is widely used as a reagent in organic chemistry especially for the synthesis of various organic compounds including pharmaceuticals and agrochemicals also Tetrabutylammonium thiocyanateIt can also be used as an electrolyte in electrochemical devices and as a phase transfer catalyst in organic synthesis Its unique chemical properties make it an important ingredient in a variety of industrial processes including petrochemical refining and materials science