Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylammonium tetrafluoroborate, For electrochemical analysis, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111014188 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4188 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4188 Supplier MilliporeSigma Supelco Supplier No. 8689625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetrabutylammonium tetrafluoroborate (TBATFB) is a quaternary ammonium salt, mainly used as electrolyte.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Weight (g/mol) 329.27
MDL Number MFCD00011634

Specifications

Melting Point 155°C to 161°C; 159°C to 162°C
CAS 429-42-5
Flash Point >93°C
Linear Formula (CH3 CH2 CH2 CH2)4 N(BF4)
Quantity 25 g
Solubility Information Methanol: 0.1 g/mL, clear, colorless
Formula Weight 329.27
Percent Purity ≥99.0% (T); ≥99.0%
Grade For electrochemical analysis
Physical Form Crystals
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.