Learn More
Medchemexpress LLC Tetrabutylammonium (trifluoromethanesulfonate) | 35895-70-6 | MFCD00042585 | 98.0% | 391.53 g·mol-1 | C17H36F3NO3S | 25 G
SDP
Catalog No. 5000208910
Encompass_Preferred
Tetrabutylammonium trifluoromethanesulfonate is a quaternary ammonium salt supplied as a solid reagent for laboratory use. It serves as a phase-transfer catalyst, a non-coordinating triflate counterion in synthesis, and an electrolyte in electrochemical and analytical applications. The material is provided with supporting documentation including a data sheet, COA, and SDS, and is intended for bench-scale research where a stable, high-purity triflate salt is required.
  • High purity solid reagent suitable for organic synthesis and analytical applications
  • Functions as a phase-transfer catalyst and non-coordinating triflate counterion
  • Available in 25 g pack size for bench-scale work
  • Store sealed at 4°C and protect from moisture
  • Documentation available: data sheet, COA, and SDS
Catalog No. 50-002-08910 Supplier Medchemexpress LLC Supplier No. 25GHYW042557
Tetrabutylammonium trifluoromethanesulfonate is a quaternary ammonium salt supplied as a solid reagent for laboratory use. It serves as a phase-transfer catalyst, a non-coordinating triflate counterion in synthesis, and an electrolyte in electrochemical and analytical applications. The material is provided with supporting documentation including a data sheet, COA, and SDS, and is intended for bench-scale research where a stable, high-purity triflate salt is required.
  • High purity solid reagent suitable for organic synthesis and analytical applications
  • Functions as a phase-transfer catalyst and non-coordinating triflate counterion
  • Available in 25 g pack size for bench-scale work
  • Store sealed at 4°C and protect from moisture
  • Documentation available: data sheet, COA, and SDS