Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetrabutylammonium trifluoromethanesulfonate | 35895-70-6 | MFCD00042585 | 98.0% | 391.53 g/mol | C17H36F3NO3S | 10 G
SDP

Catalog No. 5000210081
Encompass_Preferred

Tetrabutylammonium trifluoromethanesulfonate is a quaternary ammonium salt used as a reagent and phase-transfer catalyst in organic synthesis and biochemical assays. It provides a lipophilic tetrabutylammonium cation paired with a non-coordinating trifluoromethanesulfonate anion, enabling solubilization of ionic species in organic media.

  • Purity 98.0%.
  • Molecular weight 391.53 g/mol.
  • Chemical formula C17H36F3NO3S.
  • CAS number 35895-70-6.
  • Store at 4°C, sealed and away from moisture.
  • Available in small pack sizes for laboratory use.

Catalog No. 50-002-10081 Supplier Medchemexpress LLC Supplier No. HYW04255710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetrabutylammonium trifluoromethanesulfonate is a quaternary ammonium salt used as a reagent and phase-transfer catalyst in organic synthesis and biochemical assays. It provides a lipophilic tetrabutylammonium cation paired with a non-coordinating trifluoromethanesulfonate anion, enabling solubilization of ionic species in organic media.

  • Purity 98.0%.
  • Molecular weight 391.53 g/mol.
  • Chemical formula C17H36F3NO3S.
  • CAS number 35895-70-6.
  • Store at 4°C, sealed and away from moisture.
  • Available in small pack sizes for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.