Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrabutylphosphonium Hydroxide Solution

Catalog No. 438294100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-829-4100ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 43-829-4100ML Supplier Sigma Aldrich Supplier No. 438294100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetrabutylphosphonium hydroxide (TBPH) can be used:To convert poorly water-soluble acidic APIs into TBP ionic liquids (IL).As a cation source in the synthesis of Good'sas buffer ionic liquids (GB-ILs) via an acid-base neutralization reaction with Good'sas buffer anions.To synthesize tetrabutylphosphonium acetate, which along with Cu2O-nanoparticles forms an excellent catalyst system for protodecarboxylation reactions.

Specifications

CAS 16605-03-1
Molecular Formula C15H18NP
Refractive Index n20/D 1.412
Linear Formula C15H18NP
MDL Number MFCD00233832
Quantity 1 g
Synonym TBPH
Molecular Weight (g/mol) 243.28 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.