Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylphosphonium Tetraphenylborate 98.0+%, TCI America™
SDP

Catalog No. T200825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T200825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T200825G Supplier TCI America Supplier No. T200825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 29089-62-1
Molecular Formula C40H56BP
Molecular Weight (g/mol) 578.672
MDL Number MFCD04038112
InChI Key OGWSZNMGVXKACZ-UHFFFAOYSA-N
PubChem CID 14007839
IUPAC Name tetrabutylphosphanium;tetraphenylboranuide
SMILES [B-](C1=CC=CC=C1)(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.CCCC[P+](CCCC)(CCCC)CCCC

Specifications

Melting Point 230°C
Color White
Quantity 25 g
Formula Weight 578.67
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylphosphonium Tetraphenylborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.