Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutyltin 95.0+%, TCI America™
SDP

Catalog No. T005825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T005825G 25 g
T0058500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T005825G Supplier TCI America Supplier No. T005825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1461-25-2
Molecular Formula C16H36Sn
Molecular Weight (g/mol) 347.174
MDL Number MFCD00009422
InChI Key AFCAKJKUYFLYFK-UHFFFAOYSA-N
Synonym tetrabutyltin, tetra-n-butyltin, stannane, tetrabutyl, tin, tetrabutyl, tetra-n-butylcin, tetra-n-butylcin czech, unii-qj7y5v377v, ccris 6322, tetra-normal-butyltin, wln: 4-sn-4&4&4
PubChem CID 15098
IUPAC Name tetrabutylstannane
SMILES CCCC[Sn](CCCC)(CCCC)CCCC

Specifications

Melting Point -97°C
Color Colorless
Boiling Point 302°C
Quantity 25 g
UN Number 2788
Formula Weight 347.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetrabutyltin
EINECSNumber (2)-2270&(2)-2334
RTECSNumber WH8605000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.