Learn More
TARGETMOL CHEMICALS INC Tetrac 50MG
SDP
Catalog No. 502878789
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Tetrac (Tetraiodothyroacetic acid) is a derivative of L-thyroxine (T4), a thyroxine integrin receptor antagonist.Tetrac induces antiproliferation by blocking EGFR-mediated cell signaling in colorectal cancer cells.Tetrac blocks the effects of T4 and 3,5,3'-triiodo-L-thyronine (T3) on the cell-surface thyroxine integrin alphavbeta3 receptors.Tetra has anti-angiogenic, anti-tumor activity and pro-apoptotic activity. Purity 99.04%
Catalog No. 50-287-8789 Supplier TARGETMOL CHEMICALS INC Supplier No. T3671550MG
Also available in 1 mL, 10 mg, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Tetrac (Tetraiodothyroacetic acid) is a derivative of L-thyroxine (T4), a thyroxine integrin receptor antagonist.Tetrac induces antiproliferation by blocking EGFR-mediated cell signaling in colorectal cancer cells.Tetrac blocks the effects of T4 and 3,5,3'-triiodo-L-thyronine (T3) on the cell-surface thyroxine integrin alphavbeta3 receptors.Tetra has anti-angiogenic, anti-tumor activity and pro-apoptotic activity. Purity 99.04%