Learn More
Apexbio Technology LLC Tetracaine HCl, 100mg Cas# 136-47-0
SDP
Catalog No. 501998542
Encompass_Preferred
Tetracaine HCl is a potent local anaesthetic and a channel function allosteric inhibitor.Tetracaine hydrochloride is a calcium channel protein inhibitor and blocks voltage-sensitive release of Ca2+ from sarcoplasmic reticulum. Tetracaine is a potent local Other sizes are available. Please inqury us for quote.
Catalog No. 50-199-8542 Supplier Apexbio Technology LLC Supplier No. B1413100
Tetracaine HCl is a potent local anaesthetic and a channel function allosteric inhibitor.Tetracaine hydrochloride is a calcium channel protein inhibitor and blocks voltage-sensitive release of Ca2+ from sarcoplasmic reticulum. Tetracaine is a potent local Other sizes are available. Please inqury us for quote.