Learn More
Apexbio Technology LLC Tetracaine HCl 136-47-0 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B14135.1
Encompass_Preferred
Tetracaine hydrochloride (CAS 136-47-0) is a small molecule local anesthetic commonly used in biomedical research It acts through allosteric modulation of ion channel function specifically inhibiting calcium channels and effectively suppressing voltage-dependent calcium release from the sarcoplasmic reticulum Due to its ability to regulate calcium channel activity tetracaine HCl serves as an important investigative tool in studies exploring calcium signaling pathways muscle physiology and neuronal excitability
Catalog No. 50-000-62135