Learn More
Medchemexpress LLC Tetracaine hydrochloride | 136-47-0 | 99.91% | 1 G
Supplier: Medchemexpress LLC HYB04731G
Tetracaine hydrochloride (Amethocaine hydrochloride) is a calcium channel protein inhibitor that blocks voltage-sensitive release of Ca2+ from the sarcoplasmic reticulum. It is primarily used topically in ophthalmology and as an antipruritic. It also alters the function of calcium release channels (ryanodine receptors) that regulate calcium release from intracellular stores.
- Blocks voltage-sensitive release of Ca2+ from the sarcoplasmic reticulum.
- Used topically in ophthalmology.
- Acts as an antipruritic.
- Alters the function of calcium release channels (ryanodine receptors).
- Inhibits spontaneous calcium release events at low concentrations.
- Completely blocks calcium release at high concentrations.