Learn More
AdipoGen Tetracaine hydrochloride
SDP
Supplier:  AdipoGen CDXT0453G050
Encompass_Preferred
Chemical. CAS 136-47-0. Formula C15H24N2O2 . HCl. MW 264.4 . 36.5. Synthetic. Tetracaine is a potent local anaesthetic and a channel function allosteric inhibitor that blocks voltage-sensitive release of Ca2 from sarcoplasmic reticulum. At low concentrations, tetracaine causes an initial inhibition of spontaneous calcium release events, while at high concentrations, tetracaine blocks release completely. inhibitor of voltage-gated sodium channels such as NaV1.4 and NaV1.8. Shown also to reversibly block nicotinic acetylcholine ACh receptors nAChRs.
Catalog No. 50-255-9953