Learn More
Abcam Tetracaine hydrochloride, allosteric RyR calcium channel blocker, 25G
Catalog No. 502706129
Encompass_Preferred
MW 300.82 Da, Purity >99%. Allosteric RyR calcium channel blocker. Shows potent local anesthetic and antipruritic effects in vivo. Reversibly inhibits AChE activity (IC₅₀ = 40 μM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6129 Supplier Abcam Supplier No. AB14324225G
MW 300.82 Da, Purity >99%. Allosteric RyR calcium channel blocker. Shows potent local anesthetic and antipruritic effects in vivo. Reversibly inhibits AChE activity (IC₅₀ = 40 μM).
The product is subject to the following: Abcam Restricted Use Statement